Learn More
AG Scientific Inc X-GAL and IPTG Solution, 10 ML
Supplier: AG Scientific Inc G257010ML
Stable solution of X-Gal and IPTG. Does not contain DMSO or DMF.IPTG is combined with X-GAL to detect lac+ colonies or cells in a colorimetric assay. Distinguishes recombinants (white) from non- recombinants (blue) in cloning strategies using vectors such as Lambda-11, M13mp 18 and 19, pUC18 and 19, pUR222 containing the lacZ gene.Solution concentration: 10 mg/mL X-Gal, 8 mg/mL IPTG.Not for human therapeutic use or for medicinal purposes. For research applications only.CAS Number: 7240-90-6, 367-93-1Molecular Weight: 408.6 Da / 238.3 DaChemical Formula: C14H15BrClNO6 / C9H18O5SSolubility: WaterStorage Temperature: +4C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.