Learn More
Hach Company Wastewater Effluent Inorganics Quality Control Standard, 500 mL
Supplier: Hach Company 2833249
Wastewater Effluent Inorganics Quality Control Standard. Ready to use; no dilutions required. Mixed standard contains: Ammonia Nitrogen: 2 mg/L NH₃-N, Nitrate Nitrogen: 4 mg/L NO₃--N, COD: 25 mg/L, Phosphate: 2 mg/L PO43-. Sulfate: 50 mg/L SO₄2-. TOC: 8 mg/L. 500 mL Bottle
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.