Learn More
New England Biolabs, Inc. WarmStart Multi-Purpose LAMP/RT-LAMP 2X Master Mix (with UDG)- 100 rxn
SDP
Supplier:  New England Biolabs, Inc. M1708S
Encompass
The WarmStart Multi-Purpose LAMP/RT-LAMP 2X Master Mix (with UDG) is designed to provide a simple one-step solution for Loop-Mediated Isothermal Amplification (LAMP) of DNA or RNA (RT-LAMP) targets. LAMP and RT-LAMP are commonly used isothermal amplification techniques that provide rapid detection of a target nucleic acid using LAMP-specific primers (supplied by the user) and a strand-displacing DNA polymerase. This mix contains a blend of Bst 2.0 WarmStart DNA Polymerase and WarmStart RTx Reverse Transcriptase in an optimized buffer solution. The inclusion of thermolabile UDG prevents carryover contamination where unintended product of a previous amplification can serve as the substrate of a subsequent reaction. The WarmStart Multi-Purpose LAMP/RT-LAMP 2X Master Mix (with UDG) is compatible with multiple detection methods including turbidity detection real-time fluorescence detection and end-point visualization.
Catalog No. 50-222-9439
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.