Learn More
STEMCELL Technologies XAV939, Size: 25 mg
Supplier: STEMCELL Technologies 1001052
Inhibit tankyrase (TNKS) 1 and 2 with XAV939 (Huang et al. Nature, 2009). By inhibiting TNKS activity, XAV939 increases the protein levels of the axin-GSK3β complex and promotes the degradation of β-catenin in SW480 cells (Huang et al. Nature, 2009). This WNT pathway inhibitor inhibits colony formation of APC-deficient, β-catenin-dependent DLD-1 colorectal cancer cells (Huang et al.); induces cardiomyogenesis in mesoderm progenitor cells derived from mouse embryonic stem cells (Wang et al.); and, in combination with the SMAD inhibitors LDN193189 and SB431542, promotes induction of forebrain fates in human pluripotent stem cell lines (Maroof et al.). XAV939 is supplied as a crystalline solid with ≥ 98% purity; Molecular weight 312.3 g/mol; CAS number 284028-89-3. Visit STEMCELL.com for more information and related products.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.