Learn More
Enzo Life Sciences Wiskostatin (1 mg). CAS: 253449-04-6
Supplier: Enzo Life Sciences BMLT1260001
A selective, reversible inhibitor of N-WASP (neural Wiskott-Aldrich syndrome protein) that selectively blocks actin filament assembly. Appears to bind to N-WASP, stabilize the autoinhibited conformation and prevent the activation of Arp2/3 (actin-related protein 2/3) complex. Purity: ≥98% (HPLC). Solubility: Soluble in ethanol (10 mg/ml with warming) or DMSO (25 mg/ml). Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.