Learn More
Andwin Scientific WIPERS, STERILE 12X12 POLY CEL
SDP
Supplier:  Andwin Scientific 21914777
Encompass_Preferred
The Spec-Wipe 3 Sterile Wiper is ideal for aseptic environments where bacteria, particulate, and extractable levels are critical issues. This product is constructed of a highly absorbent 45% polyester, 55% cellulose blend that is hydroentangled to l
Catalog No. NC0341808
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.