Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Wilmad™ 3mm FEP Sample Tube Liner

Provides a contamination-free environment for NMR investigations where corrosive chemical compounds such as hydrofluoric acid, ammonium bifluoride and concentrated hydroxide solutions are present. Bel-Art SP Scienceware™ Wilmad™ 3mm FEP Sample Tube Liners are compatible with Wilmad's 7 in. NMR tubes.

Supplier:  Bel-Art™ 660000371

 View more versions of this product

Catalog No. 16-800-209


Explore all special offers
Add to Cart

Description

Description

  • Round-bottom, thin-wall construction minimizes filling-factor losses
  • NonrigidStraighten upon insertion into the sample tube.
  • PTFE plug is included with each liner.
  • Not recommended for elevated temperature studies.
Specifications

Specifications

Tube Liner
FEP
1.50 mm
6003
202 mm
1.95 mm
8 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.