Learn More
Andwin Scientific WHITE LASER TOUGH-SPOTS LARG
SDP
Supplier:  Andwin Scientific 91854000
Encompass_Preferred
The white Tough-Spots labels are specially designed to be used for 0.5 mL tubes. The labels are resistant against chemicals and temperatures from -196�C to 80�C. This product can be put on several surfaces like most plastics, glass, vinyl, and metals. Any marking instrument can also be utilized to write on these labels. This product has a diameter of 1.0".
Catalog No. 50-821-001
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.