Learn More
New England Biolabs, Inc. WarmStart Colorimetric LAMP 2X Master Mix (DNA & RNA) – 100 reactions (25 µl vol)
SDP
Supplier:  New England Biolabs, Inc. M1800S
Encompass
The WarmStart Colorimetric LAMP 2X Master Mix is an optimized formulation of Bst 2.0 WarmStart DNA Polymerase and WarmStart RTx in a special low-buffer reaction solution containing a visible pH indicator for rapid and easy detection of Loop-Mediated Isothermal Amplification (LAMP) and RT-LAMP reactions. This system is designed to provide a fast, clear visual detection of amplification based on the production of protons and subsequent drop in pH that occurs from the extensive DNA polymerase activity in a LAMP reaction, producing a change in solution color from pink to yellow. This mix can be used for any LAMP or RT-LAMP reaction and requires only a heated chamber and samples, with readout of positive amplification judged by eye in 15-40 minutes.
Catalog No. 50-152-6402
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.