Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products Half Size Human Skeletons
SCI_ED

Wired teaching skeleton for natural movement.

Supplier:  Walter Products B10202

 View more versions of this product

Catalog No. S07105


Includes: Labeled diagram
This item is not returnable. View return policy

Description

Description

  • Three-foot plastic skeleton resting on heavy metal stand
  • Removable skull with calvarium cut
  • Mandible hinged with springs in place to examine TMJ problems
  • Arms and legs held on by small screws, making it easy to bend extremities in all directions
  • All bones casted individually, except in hands and feet

Specifications

Specifications

Model, Human Skeleton with Nerve Ending
36 in.
Removable skull with the calvarium cut, mandible hinged with springs, screwed arms and legs, labeled diagram
Heavy Metal Stand
Teacher tool
91.44 cm
Plastic
Half-Size
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.