Learn More
Andwin Scientific VWR UNDERPAD BLN NS28X CS/100
SDP
Supplier:  Andwin Scientific 56616022
Encompass_Preferred
The bench underpads are made with white spunbond polypropylene facing sheet. With an absorbency of 770 mL, this product is an efficient liquid absorbent. This product also has four sealed sides, which prevent liquid extraction. It is sold in a case of 200.
Catalog No. 50-899-90308
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.