Learn More
Andwin Scientific TUBE PP NS GRD 50ML 500CS
Supplier: Andwin Scientific 21008177
The 50 mL SuperClear™ Microcentrifuge Tubes are constructed from an ultra-clear polypropylene resin that permits users to have a clear view of the pellet and accurate color rendition. This product features a labeling surface on the side that allows e