Learn More
Andwin Scientific VWR PCR 8-WELL TUBE STRIPS AN
Supplier: Andwin Scientific 53509304
The 0.2ml polypropylene PCR 8-well tube strips feature attached bubble caps and smooth walls for accurate thermal transfer. It is compatible with most popular thermal cycler blocks.