Learn More
Andwin Scientific CONTAINER 8OZ NO LID CS500
Supplier: Andwin Scientific 89107310
The Polypropylene Specimen Containers feature durability and graduations in 10 mL and 0.5 oz. increments. This product is constructed with transparent polypropylene to permit users to easily and conveniently view the contents. The container is nonste