Learn More
Andwin Scientific 96-Well PCR and Real Time PCR Plates, 200 uL, Natural, 100/cs
SDP
Supplier:  Andwin Scientific 82006636
Encompass_Preferred
Polypropylene PCR plates are compatible with most thermal cyclers and are ideal for high-throughput screening thermocycler applications. Plates have a working capacity of 200µL per well. They are thin, flexible, and easy to cut.
Catalog No. NC1266949
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.