Learn More
STEMCELL Technologies Methylcobalamin, Size: 100 mg
SDP
Supplier:  STEMCELL Technologies 1000895
Encompass_Preferred
Methylcobalamin is a vitamin B12 analog and a coenzyme involved in homocysteine metabolism (Okada et al.). Methylcobalamin elevates the activities of ERK1/2 and AKT and improves nerve regeneration in rats (Okada et al.). This small molecule has therapeutic potential for treating nervous disorders (Okada et al.).DIFFERENTIATION· Promotes Schwann cell differentiation in vitro (Nishimoto et al.).DISEASE MODELING· Promotes neuronal survival in rats (Okada et al.).
Catalog No. 50-223-1598
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.