Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scigen VIP-Fixative Leak-Proof Specimen Containers Neutral Buffered 10% Formalin Filled

Safely transport, store and retrieve pathology specimens using Scigen’s Forma-Vial™ system. Containers from 7 mL to 7.5 Liters, filled with 10% neutral buffered formalin, OSHA compliant.

Supplier:  Scigen 6128

 View more versions of this product

Catalog No. 23-730-597


Description

Description

  • Forma-Vial™ specimen containers are uniquely designed to be leak proof and protect against biohazardous or hazardous material release to the environment
  • Highly visible OSHA compliant labeling
  • Sizes range from 7mL dermatology containers to 7.5 Liters operating room organ buckets
  • Container openings range from 13mm to 300mm
  • Filled with 10% neutral buffered formalin to 50% of container’s capacity
  • Formalin waste designed to be treated in bulk using Neutralex or treated individually using Neutralexr tablets (patent pending).
  • Nonsterile containers marked with a lot code and expiration date, 4 year shelf life
  • Manufactured using ISO 13485 and FDA quality systems

Specifications

Specifications

150 mm
3800 mL
Yes
Non-sterile
Lined Induction Cap
Lined Induction Cap
Clear
Histology/cytology specimen containers
254 mm
Specimen Container Pre-filled With Neutral Buffered 10% Formalin
3800 cc
4
Specimen Container
Leak Proof Specimen Container, Vial
110 mm
3800 mL
No
PET
White
PP
128 oz.
Leakproof 3800 mL specimen container
10 in.
50% Filled Neutral Buffered 10% Formalin
OSHA Compliant
4
3800 mL (128 oz.), pre-filled 50%
128 oz.
Forma-Vial™ Specimen Containers
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.