Learn More
Cole-Parmer Monarch 6400-012 Vibration Meter with Electronic Stethoscope
Supplier:  Cole-Parmer 9883400
Encompass_Preferred
Use the vibration meter and electronic stethescope in combination with the accelerometer vibration sensor and headphones to quickly locate noise and chatter in your machinery. Vibration severity chart on front of meter provides instant status by relating the digital reading to machinery applications per ISO 10816-1. Software lets you store, compare, and graph your measurements. Software System Requirements: System: 486 computer or better, CD-ROM, running Windows 95/98, XP and NT 4.0; Memory: 8 MB RAM; Hard disk space: 100 MB minimum
Catalog No. 50-631-704
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.