Learn More
Chemglass Life Sciences Vial, 2.0mL, Clear, Glass, Shell, 12x32mm
SDP
Supplier:  Chemglass Life Sciences CV21071232
Encompass_Preferred
Vial, 2.0mL, Clear, Glass, Shell, 12x32mm. Shell vials are the most economical alternative for use in an auto sampler. Feature thicker walls for safer sample handling.Require Snap Plugs
Catalog No. 50-230-876
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.