Learn More
Chemglass Life Sciences Vial Only, Sample, 24mL, Clear, 23x85mm, GPI 20-400
Supplier: Chemglass Life Sciences CG490207
Vial Only, Sample, 24mL, Clear, 23x85mm, GPI 20-400. Clear sample vials packaged in convenient LAB-PACS. Made from Type 1 borosilicate glass. Vials are conveniently packed in shrink-wrapped packs of 100. Caps sold separately.