Learn More
Chemglass Life Sciences Vial, 2.0mL, Amber, Large Opening, 12x32mm, 11mm Crimp
Supplier: Chemglass Life Sciences CV11861232
Vial, 2.0mL, Amber, Large Opening, 12x32mm, 11mm Crimp. The 40% larger opening prevents broken needles due to the increased target area. The neck finish allows for the use of the patented Poly Crimp™ seals or standard aluminum seals. This vial is designed to work auto samples incorporating a robotic arm as well as other auto samplers. Amber borosilicate. The vial incorporates the unique Step Vial design that precisely centers a limited volume insert in the vial neck. Available with or without graduated marking spots.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.