Learn More
Chemglass Life Sciences Vial Only, Sample, 23.7mL, Clear, High Recovery, 28x67mm, GPI 22-400
SDP
Supplier:  Chemglass Life Sciences CG491605
Encompass_Preferred
Vial Only, Sample, 23.7mL, Clear, High Recovery, 28x67mm, GPI 22-400. High recovery vial with a conical bottom for complete sample recovery. These vials are the same size or slightly smaller on the OD to allow them to be interchanged with any racks or blocks that accept standard dram or scintillation vials. Packed in an SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identification. Vial only.
Catalog No. 50-230-672
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.