Learn More
Chemglass Life Sciences Vial, 6.0mL, Clear, 20mm Headspace, Flat Bottom, 22x38mm, 20mm Crimp
Supplier: Chemglass Life Sciences CV12802238
Vial, 6.0mL, Clear, 20mm Headspace, Flat Bottom, 22x38mm, 20mm Crimp. These vials are manufactured from Type 1 Class A Borosilicate glass. The standard flat bottom vials can be used with Hewlett-Packard, Carlo Erba and Shimadzu. The European 20mm aluminum seal vials with rounded bottoms can be used in Perkin-Elmer, Tekmar, CTC/L.E.A.P.™ Technologies, Gertsel and Varian.Please Note: Pressure Release Aluminum Seals are Designed to Open When the Internal Pressure Exceeds 3.0 +/- 0.5 Bar
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.