Learn More
TWD Scientific LLC BioX 500uL SBS Format Minitube Combo. Case of 1920 Tubes and 20 Racks.
SDP
Supplier:  TWD Scientific LLC BX894253
Encompass_Preferred
BioX 500µL SBS format screw top minitubes with blue caps. Vials come loaded into a barcoded 96-well rack with square wells to eliminate slip. The internally threaded crovials feature 2D barcodes on the bottom and are gamma radiation sterilized. 96 tubes/rack, 20 racks/case. price/case.
Catalog No. 50-227-4264
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.