Learn More
Chemglass Life Sciences Vial, 20mL, Amber, 18mm Screw Thread Headspace, Flat Bottom, 22x75mm
SDP
Supplier:  Chemglass Life Sciences CV12942275
Encompass_Preferred
Vial, 20mL, Amber, 18mm Screw Thread Headspace, Flat Bottom, 22x75mm. These vials are manufactured from Type 1 Class A borosilicate glass. The radius bottom vials can be used on LEAP Technologies Inc, CTC Analytics, Varian, Gerstel and other instruments using CombiPAL and other models requiring magnetic capable closures.
Catalog No. 50-230-795
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.