Learn More
Chemglass Life Sciences Vial, 2.0mL, RAM™, Clear with White Grad Spot, Large Opening, 12x32mm, 9mm Thread
Supplier: Chemglass Life Sciences CV11521232
Vial, 2.0mL, RAM™, Clear with White Grad Spot, Large Opening, 12x32mm, 9mm Thread. With a 40% larger opening, the RAM™ Vial is specifically designed to work in robotic arm auto samplers. These vials incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial. Available with graduated marking spots, and silanized.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.