Learn More
Chemglass Life Sciences Vial, 2.0mL, Amber, Large Opening, 12x32mm, 9mm Thread
SDP
Supplier:  Chemglass Life Sciences CV11511232
Encompass_Preferred
Vial, 2.0mL, Amber, Large Opening, 12x32mm, 9mm Thread. With a 40% larger opening, these vials are specifically designed to work in robotic arm auto samplers. They also incorporate the unique Step Vial design that precisely centers a limited volume insert in the neck of the vial. Available with graduated marking spots, and silanized.
Catalog No. 50-230-736
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.