Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STERIS™ Vesta Syde™ SQ 64 st RTU Disinfectant
Sterile version of phosphate-free one-step cleaner and disinfectant, formulated for use on hard, non-porous surfaces
Supplier: STERIS™ 1S24Y2
This item is not returnable.
View return policy
Description
- Kills broad spectrum of common bacteria and fungi
- Use-dilution rate of 01.6 % v/v (1:64)
- Requires no mixing or activation for convenient and consistent use
- Specifically formulated for disinfection of hard surfaces
- Can be applied manually or sprayed
- Ease of Use- Eliminates need to validate dilution preparation procedures by providing consistent dilution every time
- Convenience- Same key ingredients as Vesta-Syde™ SQ Disinfectant concentrate to simplify validation requirements
- Productivity- Low residue formulation to maintain visibly clean surfaces with minimal rinsing
- Efficacy- Microbial control against bacteria, fungi, and viruses using AOAC and EN protocols
- Availability- Active ingredients notified according to Biocide Product Directive and registered with US EPA
- Assurance- Manufactured in accordance with cGMP with complete lot traceability
Specifications
Quaternary Disinfectant | |
Sterile | |
12/Cs. |
10.9 | |
Quaternary Ammonium | |
23 oz. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction