Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Mixed Anion Standard 7, 150ppm SO42-, PO43-; 100ppm NO3-, Br-, NO2-; 30ppm Cl-, 20ppm F-; ISO 17025/Guide 34, Ricca Chemical
SDP

Formulated for high performance and reliable results

Supplier:  Ricca Chemical Company RV010685100N

 View more versions of this product

Catalog No. RV010685100


Explore all special offers
Add to Cart

Description

Description

  • 150ppm SO42-, PO43; 100ppm NO3-, Br-, NO2-; 30ppm Cl-, 20ppm F-
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

0.98%, 0.1176%, 0.0196%, 0.0098%, 0.004802%
7722-76-1, 7783-20-2, 7631-99-4, 7647-14-5, 7732-18-5
High Purity
100mL
1.02%, 0.1224%, 0.0204%, 0.0102%, 0.004998%
Liquid
Bottle, Poly Natural
1.07
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.