Learn More
Vector Laboratories Normal Goat Serum 2.5% AK-6000
SDP
Supplier:  Vector Laboratories AK6000
Encompass_Preferred
Normal Goat Serum Blocking Solution, 2.5%, 50 ml, is ready-to-use and is prepared identically to Vector Laboratories undiluted normal serum. The pooled sample collected from healthy adult animals is heat-treated and centrifuged to remove precipitates and then filtered. The serum is tested with the appropriate antibody to check for possible cross-reactivities. The serum is then diluted to 2.5% (v/v) in buffer containing 0.08% sodium azide. This is the same serum preparation that is included in ImmPRESS Detection Systems. This reagent can be used for blocking non-specific binding or as an antibody diluent.
Catalog No. 50-204-5598
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.