Learn More
Enzo Life Sciences [pSer239]VASP monoclonal antibody (16C2) (FITC conjugate) (100 µg)
SDP
Supplier:  Enzo Life Sciences ALX804240FC100
Encompass_Preferred
Synonyms: Vasodilator stimulated phosphoprotein. Clone: 16C2. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of VASP (vasodilator stimulated phosphoprotein) phosphorylated at Ser239 (RKVpSKQE) conjugated to KLH. UniProt ID: P50552. Species reactivity: Human, Mouse, Rat, Rabbit. Applications: Flow Cytometry, ICC. Purification: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Liquid. 0.5mg/ml in 2x PBS containing 0.09% sodium azide, PEG and sucrose.
Catalog No. 50-210-0507
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.