Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Vanderzant-Adkisson Special Wheat Germ Diet for Insects

Vanderzant-Adkisson Special Wheat Germ Diet for Insects is widely used for laboratory rearing of more than 20 species of insects.

Specifications

Product Type Insect Diet
Format Powder
View More Specs

Products 3
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
02-902-94210
View Documents
MP Biomedicals™
0290294210
10 kg N/A N/A N/A
02-902-94220
View Documents
MP Biomedicals™
0290294220
20 kg N/A N/A N/A
02-902-94250
View Documents
MP Biomedicals™
0290294250
50 kg N/A N/A N/A
Description

Description

  • Vanderzant-Adkisson Special Wheat Germ Diet for Insects is a special wheat germ diet for the rearing of various insects.
  • Vanderzant-Adkisson modification is widely used for laboratory rearing of more than 20 species of insects.
  • Included among these are such important species of the order Lepidoptera as the corn earworm (Heliothis zea), the tobacco budworm (Heliothis virescens), the pink bollworm (Pectinophora gossypiella), the cabbage looper (Trichoplusiani), beet armyworm (Spodoptera exigua), yellow-striped armyworm (Prodenia ornithogalli), European corn borer (Ostrinia nubilalis), sugarcane borer (Diatraea saccharalis), and codling moth (Carpocapsa pomonella) and others.
  • Also included are two species of Curculionidae, the boll weevil (Anthonomus grandis) and the alpha weevil (Hypera postica).
  • The Special Wheat Germ Diet, when used with Vanderzant's Vitamin Fortification Mixture, has been very successful for rearing the corn earworm, the tobacco budworm and the pink bollworm.
Specifications

Specifications

Insect Diet
Powder
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.