Learn More
Gold Biotechnology Inc Vancomycin Hydrochloride, USP Grade, 1404-93-9, MFCD03613611, 25 g
SDP
Supplier:  Gold Biotechnology Inc V20025
Encompass_Preferred
Formula: C 66H75Cl2N9O24 HCl, MW: 1485.71 g/mol, Store desiccated at -20°C, Soluble in water, Potency of greater than 1000 µg/mg. Vancomycin hydrochloride is a member of the glycopeptide antibiotic class derived from Streptomyces orientalis. It is effective against gram-positive bacteria and used in studies of antibiotic resistance bacteria and nanoparticle transport studies.
Catalog No. 50-153-2847
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.