Learn More
Electron Microscopy Sciences Vacuum Oven Model 3618-6, 240 V, 14"W x 14"H x 20"D chamber, 2.3 cu. ft (65 L) Capacity
Supplier: Electron Microscopy Sciences 6323820
Multipurpose vacuum oven with 220 deree C maximum temperature conserves chamber space for drying, curing, vacuum embedding, and plating applications. Silicone door gasket and a positive latch door maintains the seal for all vacuum levels. Exclusive Polycarbonate safety shield protects door glass. Two removable aluminum shelves including standard. Easy-to-clean #304 staniless steel chamber interior. Powder coated heavy guage steel exterior construction.
Chamber: 14"W x 14"H x 20"D
Capacity: 2.3 cu. ft (65 L)
Thermometer: LED
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.