Learn More
Proteintech Group Inc V5-Trap Agarose
SDP
Supplier:  Proteintech Group Inc V5TA20
Encompass
V5-Trap Agarose is an affinity reagent for IP and purification of V5-tagged proteins. It consists of an anti-V5 Nanobody/ VHH coupled to agarose beads.
Catalog No. 50-225-8896
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.