Learn More
Medipak UVLI Slit-Top Covers 0861, Color- Black, Standard Size- 8 in x 14 in, 500 Bags
Supplier:  Medipak 0861
Encompass_Preferred
For 1-liter IV bags (1000 ml). Black UVLI™ covers provide a convenient, cost-effective, and disposable solution for protecting light-sensitive medications during transportation and administration. Our Black UVLI™ covers block 99% or more of light transmission and are opaque, making them the perfect solution for blinding. Slit-top UVLI™ covers are fully sealed on two sides with the top seal having a convenient unsealed portion for even easier hanging.
Catalog No. NC3357140
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.