Learn More
Copan Diagnostics™ UriSponge™: Urine Specimen Collection, Preservation and Transport System
SDP
Allows safe collection and preservation of urine specimens without the added expense and hazard of hypodermic urine transfer straws
Supplier:  Copan Diagnostics™ 8C020S100.A
 View more versions of this product
Catalog No. 23-600-528
Description
Description
  • Urine collection device combines a plastic tube and a screw cap with attached sponges impregnated with boric acid and sodium formate urine preservatives
  • Maintains viability of bacteria at refrigerated and room temperature for up to 48 hours
  • No hypodermic needle urine transfer straw needed, eliminating potential injury and sharps disposal costs
  • Reduce repeat collections from patient due to inaccurate filling of boric acid tubes
  • Automatically processed on WASP™ using its built-in spinner, which easily elutes urine from the sponges prior to planting and streaking
  • If processed manually, simply centrifuge the tube to release urine from the sponge
Specifications
Specifications
Foam Sponge Urine Collection Device
Polypropylene with Polyurethane Foam Sponge
Bulk
Screw Cap
500/Cs.
16 x 100 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.