Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Universal Lateral Flow Assay Kit
The Expedeon Universal Lateral Flow Assay (LFA) Kit is designed to enable the easy development of customized sandwich lateral flow assays.
Supplier: Novus Biologicals™ 43000100
Description
Kit Contents:
- 3 x 100ug Lightning-Link™ Ulfa-Tag conjugation kit
- 3 x MINI reaction 40 nm InnovaCoat™ GOLD
- 40 nm InnovaCoat GOLD™-Biotin
- 2 x 96-well low binding well plates
- 100 Universal-LFA strips
- Buffer
- Scoring card
Specifications
Assay Kit | |
100 Tests | |
Storage of components varies. See protocol for specific instructions. | |
Universal Lateral Flow |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction