Learn More
Great Lakes Diagnostics™ Universal Fit Pipette Tip
Great Lakes Diagnostics™ universal fit pipette tips are available in different volumes and lengths. Compatible with a broad range of pipettes.
Supplier: Great Lakes Diagnostics™ FT682201
Description
- Sterile
- Filtered
- Racked
- DNA/RNA free
- Max Recovery
- Compatible with:
- Eppendorf Reference™ SC, vol .5-10 μL, 2-20 μL, 10-100 μL, 50-200 μL, 100-1000 μL
- Thermo Finnpipette™ F1/F2 SC, 1-10 μL, 2-20 μL, 10-100 μL, 20-200 μL, 100-1000 μL
- Finnpipette F3 SC 1-10 μL, 10-100 μL
- Biohit Proline™ SC .5-10 μL, 20 μL, 5-50 μL, 20-200 μL, 100-1000 μL
- Gilson Pipetman™ SC .5-10 μL, 20 μL, 5-50 μL, 20-200 μL, 100-1000 μL
- Axygen™ SC and MC pipettes.
- Corning™ pipettes.
Specifications
20 μL | |
Sterile | |
Autoclavable | |
Clear | |
Axygen™ SC and MC, Biohit™ Proline™ SC, Corning™, Eppendorf Reference, Gilson™ Pipetman™ SC, Axygen™ SC and MC, Corning™ |
Filtered | |
Clear | |
Rack | |
Single-use | |
No |