Learn More
Glass Expansion UniFit with 2.0mm OD x 0.86mm ID x 700mm long tube (PKT 3)
Supplier: Glass Expansion UFT2700
UniFit with 2.0mm OD x 0.86mm ID x 700mm long tube (PKT 3)Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001