Learn More
Andwin Scientific UNDERPADS--23INX24 200 CASE
Supplier: Andwin Scientific 56616032
The Deluxe Absorbent Underpads have a three-layer, cushioned construction that is breathable yet absorbent enough. This product is ideal for hospital and laboratory use. The top sheet is made with soft white spunbound polypropylene, while its back sh