Learn More
Electron Microscopy Sciences Branson Ultra Sonic Bath Model 1800-MT, 115 V
Supplier: Electron Microscopy Sciences 71020MT
With a unique sweep frequency technology to eliminate standing waves, & a pioneering 40 kHz industrial transducers, Bransonic ultrasonic baths have been the industry standard for quality, reliability, & precision cleaning.Constant activity/power tracking automatically maintains the ultrasonic power by adjusting for changes in liquid level & temperature caused by a light or heavy load. This helps ensure more uniform cleaning & consistent performance.High/low power control adjusts the acoustic energy in the bath & can be set at 100% power for normal to heavier loads or at 70% power for lighter, This helps to protect components from potential damage.Temperature settings up to 69°C/156.2°F can be set in Celsius or Fahrenheit & programmed through the front panel for easy access & monitoring. High-temperature visual alarm/auto shut-off provides added safety. Ultrasonic cleaner only w/ mechanical timer, tank size: 5.5inx6inx4in, tank capacity: 0.5 gal (1.8L), overall size: 10inx12inx11.5in.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.