Learn More
Research Products International Corp Ultra Clear Cellophane, 26x32cm
Supplier: Research Products International Corp 1091
The membrane used to air dry gels is the most important component. This ultra clear cellophane provides the optimum porosity to ensure uniform drying and to prevent cRKing of gels.Outperforms conventional PLAS filmLaboratory tested for gel dryingConsistant porosity and quality Pre-Cut Sheets 26 x 32 cm for Sequencing Gel DryerShipping Weight: 1 lbsShipping Dimensions: 18.00 x 7.00 x 1.00Product usually ships within 24 hours.Research or further manufacturing use only, not for food or drug use.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.