Learn More
Apexbio Technology LLC DiscoveryProbe™ Ubiquitination Compound Library, 250uL/well(10 mM solution),96 Well Racks With Screw Cap
Supplier: Apexbio Technology LLC L1049.25
DiscoveryProbe™ Ubiquitination Compound Library is a collection of 144 ubiquitination compounds supplied as lyophilized powder or pre-dissolved DMSO solutions. Available in stock with overnight delivery and free shipping over $500 Cost-effective and competitive price to save your findings Potent, selective and cell-permeable in inhibiting or activating target molecules Diverse in chemical structure and route of administration (oral/i.m/i.v injection etc.) Detailed files describing potency, selectivity and applications etc. Supported by published data from top peer-reviewed journals Guaranteed high quality with NMR and HPLC validation. Other sizes are also available. Please inqury us for quote.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.