Learn More
Enzo Life Sciences Ubiquitin (bovine), (native) (methylated) (1 mg)
SDP
Supplier:  Enzo Life Sciences BMLUW85550001
Encompass_Preferred
Octa-dimethyl-ubiquitin: (CH3)2-MQIFVK*TLTGK*TITLEVEPSDTIENV K*AK*IQDK*EGIPPDQQRLIFAGK*QLEDGR TLSDYNIQK*ESTLHLVLRLRGG-OH where K* = (ε-dimethyl)lysyl. Octa-dimethyl-ubiquitin: (CH3)2-MQIFVK*TLTGK*TITLEVEPSDTIENV K*AK*IQDK*EGIPPDQQRLIFAGK*QLEDGR TLSDYNIQK*ESTLHLVLRLRGG-OH where K* = (ε-dimethyl)lysyl. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized in HPLC-grade water. Source: Native bovine. Solubility: Soluble and stable in DMSO and aqueous buffers up to 100 mg/mL. The solid will dissolve with gentle vortexing at the desired concentration. Long Term Storage: -20°C.
Catalog No. 50-201-2285
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.