Learn More
Enzo Life Sciences U-73343 (5 mg). CAS: 142878-12-4
Supplier: Enzo Life Sciences BMLST3920005
Inactive analog of U-73122 (BML-ST391) which only slightly inhibits phospholipase C. May be used as a negative control. Alternative name: 1-[6-[[(17beta)-3-Methoxyestra-1,3,5(10)-trien-17-yl]amino]hexyl]-pyrrolidine-2,5-dione. Purity: ≥98% (HPLC). Solubility: Soluble in methylene chloride or chloroform; slightly soluble in DMSO (2mg/ml) or 100% ethanol (1mg/ml); sparingly soluble in aqueous solutions. Long Term Storage: Ambient.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.