Learn More
Enzo Life Sciences U-73122 (25 mg). CAS: 112648-68-7
Supplier: Enzo Life Sciences BMLST3910025
Inhibits agonist induced phospholipase C (PLC) activation (IC50=1-2.1 µM) in human platelets. It also inhibits agonist induced down-regulation of muscarinic receptors in SK-N-SH neuroblastoma cells. It is a useful tool to investigate receptor-mediated PI turnover in signal transduction. It is a potent inhibitor of human neutrophil adhesion to biological surfaces as well as adhesion-dependent granule exocytosis and oxidative burst. Inhibits 5-lipoxygenase in vivo and in vitro (IC50 = 30nM for recombinant enzyme and 2.4µM for PMNL homogenates). Alternative name: 1-(6-((17beta-3-methoxyestra-1 3 5(10)-trien-17-yl)amino)hexyl)-1H-pyrrole-2 5-dione. Purity: ≥98% (HPLC). Solubility: Soluble in methylene chloride or chloroform. Slightly soluble in DMSO (2mg/ml) or 100% ethanol (1mg/ml), sparingly soluble in aqueous solutions. . Long Term Storage: Ambient.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.