Learn More
Enzo Life Sciences Tyrphostin 25 (RG-50875) (5mg). CAS: 118409-58-8
SDP
Supplier:  Enzo Life Sciences BMLEI1870005
Encompass_Preferred
A potent inhibitor of EGF receptor kinase activity (IC50=3 µM). Purity: ≥99%. Solubility: Soluble in 100% ethanol or DMSO. Long Term Storage: Ambient.
Catalog No. 50-201-0594
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.