Learn More
Saint Gobain Performance Plastics Tube Tygon 3/16 x 1/16 50FT-15M/PK
Supplier:  Saint Gobain Performance Plastics AAG00012
Encompass_Preferred
Saint-Gobain Tygon Fuel and Lubricant Tubing Formula F-4040-A Diameter Outer 5/16 in. Maximum Pressure 35 psi Thickness 0.0625 in. Diameter Inner 0.1875 in. Max. Operating Temperature 74 deg.C Min. Operating Temperature -37 deg.C Overall Length 50 ft. Yellow
Catalog No. 50-206-8926
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.