Learn More
Saint Gobain Performance Plastics Tube Tygon 1/4 x 1/2 50FT-15M/PK
Supplier: Saint Gobain Performance Plastics ACF00019F
Saint-Gobain Tygon S3 E-3603 Flexible Tubings Diameter Inner 1/4 in. Diameter Outer 1/2 in. Thickness 1/8 in. Overall Length 50 ft. 15 m Clear